Cole-Parmer® Cylindrical Cuvettes with PTFE Resin Stoppers
Easily view samples in clear cuvettes
  • Cylindrical Cuvettes are perfect for applications requiring large sample windows and volumes
  • Cylindrical Cuvettes include two stoppers of PTFE resin
Choose from glass or quartz cuvette materials
28,000.00 - ₹92,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Capacity (mL) Pathlength (mm)
Availability Pricing
COMPARE
Glass 14.1 50
₹29,000.00INR / Each
COMPARE
Glass 28.2 100
₹28,000.00INR / Each
COMPARE
Quartz 14.1 50
₹78,200.00INR / Each
COMPARE
Quartz 28.2 100
₹92,900.00INR / Each