Cole-Parmer® Cooling Transport Boxes
Transport your critical samples without worry
  • Maintain specific temperature ranges for extended periods of time for improved sample protection
  • Get the temperature protection you need with choice of durable transport bags or boxes
  • Modular, color-coded cooling packs allow flexible configurations for different-sized samples
  • Ensure sample integrity with optional security sealing ties
Durable polypropylene-insulated walls offer improved insulation for better sample protection. Organize cooling packs to suit your sample or choose from one of our prepared kits. Cooling packs can be added so that no weight is placed on top of your sample (see box images). Boxes are stackable and feature a convenient shoulder strap and integrated handle.
92,100.00 - ₹169,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Volume (Liters) Description
Availability Pricing
COMPARE
290 x 170 x 130 mm 5 Transport Box, 5 L
₹97,400.00INR / Each
COMPARE
290 x 170 x 250 mm 10 Transport Box, 10 L
₹118,000.00INR / Each
COMPARE
321 x 285 x 230 mm 20 Transport Box, 20 L
₹141,000.00INR / Each
COMPARE
290 x 170 x 130 mm 5 Transport Box 5 L with Two 4°C End-Caps and Two 4°C Frames
₹92,100.00INR / Each
COMPARE
290 x 170 x 130 mm 5 Transport Box 5 L with Two 22°C End-Caps and Two 22°C Frames
₹92,100.00INR / Each
COMPARE
290 x 170 x 250 mm 10 Transport Box 10 L with Two 4°C End-Caps and Four 4°C Frames
₹126,000.00INR / Each
COMPARE
290 x 170 x 250 mm 10 Transport Box 10 L with Two 22°C End-Caps and Four 22°C Frames
₹126,000.00INR / Each
COMPARE
321 x 285 x 230 mm 20 Transport Box 20 L with Two 4°C Blocks (2 L)
₹99,900.00INR / Each
COMPARE
321 x 285 x 230 mm 20 Transport Box 20 L with Two 22°C Blocks (2 L)
₹169,000.00INR / Each
Showing 1- 9 of 9