Cole-Parmer® Concavity Microscope Slides, Glass
Enables dry-mounting of large specimens directly on slides
  • Constructed of durable sheet glass with ground edges
  • Spherical concavities are approximately 16mm in diameter and 0.8mm deep
  • Slides measure 75 x 25 mm
These Cole-Parmer® Essentials concavity microscope slides feature spherical concavities that enable dry-mounting of large specimens and are ideal for use with specimens that are too thick for conventional slides. Pre-cleaned slides feature ground edges and concavities that are 16 mm in diameter and 0.8 mm deep.
Choose from standard slides that are 1.3 mm thick or thick slides that are 3 mm thick to provide enhanced durability.
1,900.00 - ₹4,100.00INR / Pkg of 12
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Slide Type Thickness
Availability Pricing
COMPARE
1 Well 1.3 mm
₹1,900.00INR / Pkg of 12
COMPARE
2 Wells 1.3 mm
₹1,900.00INR / Pkg of 12
COMPARE
3 Wells 1.3 mm
₹3,500.00INR / Pkg of 12
COMPARE
1 Well 3 mm
₹2,300.00INR / Pkg of 12
COMPARE
2 Wells 3 mm
₹2,900.00INR / Pkg of 12
COMPARE
2 Wells 3 mm
₹4,100.00INR / Pkg of 12