Cole-Parmer® Column Temperature Control Systems
Choose from single, programmable, or high-capacity units to suit your application needs
  • Accurately control column temperature with PID temperature control
These Cole-Parmer® column temperature control systems provide even chamber temperatures.
278,000.00 - ₹672,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
90 25 90 to 260
₹278,000.00INR / Each
COMPARE
70 4 90 to 260
Call For Price
COMPARE
100 4 90 to 260
₹672,000.00INR / Each