Cole-Parmer® Coated Wire Tube Racks
Slotted bottom offers additional support for tubes and bottles
  • Chemical resistant
  • Withstands temperatures down to -40 °C
  • Autoclavable at 121 °C
The HDPE coated wire racks are chemical resistant and durable for transport or sample handling. Square wells and three deck levels help keep tubes secure while a slotted bottom provides additional support. Holds all brands and types of conical, round or flat bottom, glass
or plastic, tubes and/or bottles.
4,500.00 - ₹6,800.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Tube Size (mm) Min Tube Size (mm) Tubes Held
Availability Pricing
COMPARE
13 10 72
₹4,800.00INR / Each
COMPARE
13 10 108
₹4,800.00INR / Each
COMPARE
16 13 72
₹4,800.00INR / Each
COMPARE
16 13 108
₹4,800.00INR / Each
COMPARE
20 16 15
₹4,900.00INR / Each
COMPARE
30 25 8
₹4,500.00INR / Each
COMPARE
30 25 48
₹6,800.00INR / Each
Showing 1- 7 of 7