Cole-Parmer® Cleanroom Hose Barb Fittings, Straight Union, Natural Kynar®
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • Biopharma compatible and USP 661 and BPOG compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily connect two tubing with straight unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
1,500.00 - ₹9,100.00INR / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
3/32 3/32 Natural Kynar®
₹1,500.00INR / Pkg of 10
COMPARE
5/8 5/8 Natural Kynar®
₹5,700.00INR / Pkg of 10
COMPARE
1/16 1/16 Natural Kynar®
₹1,500.00INR / Pkg of 10
COMPARE
3/4 3/4 Natural Kynar®
₹7,100.00INR / Pkg of 10
COMPARE
1 1 Natural Kynar®
₹9,100.00INR / Pkg of 10
COMPARE
5/32 5/32 Natural Kynar®
₹2,800.00INR / Pkg of 10
COMPARE
1/8 1/8 Natural Kynar®
₹1,600.00INR / Pkg of 10
COMPARE
3/16 3/16 Natural Kynar®
₹3,100.00INR / Pkg of 10
COMPARE
1/4 1/4 Natural Kynar®
₹3,700.00INR / Pkg of 10
COMPARE
5/16 5/16 Natural Kynar®
₹4,000.00INR / Pkg of 10
COMPARE
3/8 3/8 Natural Kynar®
₹4,400.00INR / Pkg of 10
COMPARE
1/2 1/2 Natural Kynar®
₹5,000.00INR / Pkg of 10
Showing 1- 12 of 12