Cole-Parmer® Chempette® Macropipettors
Lock volume in place with set screw
Quickly adjust volume by turning the thumb knob until the yellow sight line corresponds to the desired setting on the volume window. Tighten a brass set screw, and your volume setting is locked in place.
Polypropylene construction and durable design reduce friction wear and increase precision. A piston of PTFE ensures complete sealing in a polypropylene cylinder. O-rings are not exposed to friction wear. You can draw and deliver precise samples for the entire life of the piston and cylinder.
49,900.00 - ₹54,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Min Volume (mL)
Availability Pricing
COMPARE
1 0.2
₹50,100.00INR / Each
COMPARE
2 0.4
₹49,900.00INR / Each
COMPARE
5 1
₹54,300.00INR / Each
COMPARE
10 2
₹50,400.00INR / Each