Cole-Parmer® Canine Infection Detection Kits for qPCR
Quickly and accurately detect the presence of canine infections
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
132,000.00 - ₹352,000.00INR / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Ehrlichia canis No DNA
₹161,000.00INR / Each
COMPARE
Ehrlichia canis Yes DNA
₹206,000.00INR / Each
COMPARE
Mycoplasma haemofelis and M. haemocanis No DNA
₹161,000.00INR / Each
COMPARE
Neospora caninum No DNA
₹161,000.00INR / Each
COMPARE
Neospora caninum Yes DNA
₹206,000.00INR / Each
COMPARE
Canine Adenovirus 1 No DNA
₹161,000.00INR / Each
COMPARE
Canine Adenovirus 1 Yes DNA
₹206,000.00INR / Each
COMPARE
Canine Adenovirus 2 No DNA
₹161,000.00INR / Each
COMPARE
Canine Adenovirus 2 Yes DNA
₹206,000.00INR / Each
COMPARE
No RNA
₹159,000.00INR / Each
COMPARE
Canine Coronavirus Yes RNA
₹132,000.00INR / Each
COMPARE
Canine Distemper Virus No RNA
₹161,000.00INR / Each
COMPARE
Canine Distemper Virus Yes RNA
₹206,000.00INR / Each
COMPARE
Canine Herpes Virus No DNA
₹161,000.00INR / Each
COMPARE
Canine Herpes Virus Yes DNA
₹206,000.00INR / Each
COMPARE
Canine Norovirus No RNA
₹161,000.00INR / Each
COMPARE
Canine Norovirus Yes RNA
₹206,000.00INR / Each
COMPARE
Canine Parainfluenza Virus No RNA
₹161,000.00INR / Each
COMPARE
Canine Parainfluenza Virus Yes RNA
₹206,000.00INR / Each
COMPARE
No DNA
₹161,000.00INR / Each
Showing 1 - 20 of 21