Cole-Parmer® Cable Zip Ties, Color Nylon
Color options for easy bundle identification
  • Holds securely—even in vibrating, oily, or wet environments
  • Won’t corrode or stretch
  • Withstands temperatures to 85 °C (185 °F)
  • Available in tensile strengths of 18 or 40 lbs
Available in five colors or assorted color packs, these high-quality 6/6 nylon cable zip ties provide a lasting solution for organization of your wire and cable bundles. They have a durable locking mechanism that holds securely, even when used to bundle vibrating or flexing materials. They will not corrode or stretch and will continue to hold tension even in oily spray or wet environments. A bent tail allows for easy pickup of the tie and threading through the lock.
Able to withstand temperatures to 85 °C (185 °F), these cable/zip ties have a melting point of 257 °C (495 °F) and remain flexible to -40 °C (-40 °F). Efficient range of ties may be higher or lower in your specific application. They are RoHS compliant, UL recognized, and UL 94 V-2 flammability rated.
Easily install by wrapping the tie around the bundle and threading the tail through the locking mechanism. Pull tight by hand and cut the tail to complete or use a tensioning tool—available separately. For mounting wires and cables to walls or other surfaces, use Cole-Parmer® Self Adhering Cable Zip Tie Mounts—available separately.
300.00 - ₹23,500.00INR / Pkg of 1000
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Tensile Strength (lbs)
Availability Pricing
COMPARE
Red 7 29/32 40
₹23,500.00INR / Pkg of 1000
COMPARE
Blue 7 29/32 40
₹13,400.00INR / Pkg of 1000
COMPARE
Green 7 29/32 40
₹23,500.00INR / Pkg of 1000
COMPARE
Orange 7 29/32 40
₹19,700.00INR / Pkg of 1000
COMPARE
Red 4 18
₹9,600.00INR / Pkg of 1000
COMPARE
Blue 4 18
₹9,600.00INR / Pkg of 1000
COMPARE
Green 4 18
₹9,600.00INR / Pkg of 1000
COMPARE
Yellow 4 18
₹6,600.00INR / Pkg of 1000
COMPARE
Orange 4 18
₹9,600.00INR / Pkg of 1000
COMPARE
Red 4 18
₹300.00INR / Case of 100
COMPARE
Assorted 4 18
₹4,400.00INR / Case of 700
COMPARE
Green 8 40
₹1,000.00INR / Case of 100
COMPARE
Yellow 8 40
₹6,000.00INR / Case of 1000
COMPARE
Assorted 8 40
₹9,700.00INR / Case of 700
Showing 1- 14 of 14