Cole-Parmer® Brix Analog Refractometers
Quick and easy operation with reliable measurements
  • Easy calibration via the calibration screw for enhanced accuracy
  • No more worrying about temperature fluctuations with the automatic temperature compensation (ATC) models
These units come in multiple brixrange options, including models with multiple brix scales. For the units that feature automatic temperature compensation (ATC), these allow the user to concentrate on the measurements without worrying about the temperature. The compensation range is 50 to 86°F (10 to 30°C).
All models also feature a zero% adjust with distilled water. This allows for immediate and easy calibration for precise measurements at all times.
18,500.00 - ₹51,600.00INR / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Brix Range (%) Min Brix Range (%) Resolution
Availability Pricing
COMPARE
10 0 0.001
₹18,500.00INR / Each
COMPARE
18 0 0.001
₹20,400.00INR / Each
COMPARE
32 0 0.2%
₹18,500.00INR / Each
COMPARE
62 28 0.002
₹21,800.00INR / Each
COMPARE
82 45 0.005
₹20,300.00INR / Each
COMPARE
92 58 0.002
₹21,000.00INR / Each
COMPARE
60 0 0.005
₹18,800.00INR / Each
COMPARE
80 0 0.005
₹27,800.00INR / Pkg of 1
COMPARE
80 0 0.005
₹20,200.00INR / Pkg of 1
COMPARE
90 0 0.005
₹18,600.00INR / Each
COMPARE
90 0 0.002
₹51,600.00INR / Each
COMPARE
10 0 0.001
₹18,500.00INR / Each
COMPARE
18 0 0.001
₹18,500.00INR / Each
COMPARE
32 0 0.002
₹20,500.00INR / Each
COMPARE
62 28 0.002
₹21,000.00INR / Each
COMPARE
82 45 0.005
₹22,400.00INR / Each
COMPARE
92 58 0.002
₹20,600.00INR / Each
Showing 1- 17 of 17