Cole-Parmer® Bovine Infection Detection Kits for qPCR
Quickly and accurately detect the presence of bovine infections
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
155,000.00 - ₹273,000.00INR / Each
23 variations of this product are available.
See All Product Options Below
23 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Encephalitozoon (All Species) No DNA
₹206,000.00INR / Each
COMPARE
Brucella abortus No DNA
₹161,000.00INR / Each
COMPARE
Brucella abortus Yes DNA
₹206,000.00INR / Each
COMPARE
Campylobacter fetus No DNA
₹161,000.00INR / Each
COMPARE
Campylobacter fetus Yes DNA
₹206,000.00INR / Each
COMPARE
Chlamydophila abortus No DNA
₹161,000.00INR / Each
COMPARE
Chlamydophila abortus Yes DNA
₹206,000.00INR / Each
COMPARE
Mycoplasma bovis No DNA
₹161,000.00INR / Each
COMPARE
Mycoplasma bovis Yes DNA
₹206,000.00INR / Each
COMPARE
Streptococcus agalactiae No DNA
₹161,000.00INR / Each
COMPARE
Streptococcus agalactiae Yes DNA
₹206,000.00INR / Each
COMPARE
Theileria parva No DNA
₹161,000.00INR / Each
COMPARE
Theileria parva Yes DNA
₹206,000.00INR / Each
COMPARE
Tritrichomonas foetus No DNA
₹161,000.00INR / Each
COMPARE
Tritrichomonas foetus Yes DNA
₹206,000.00INR / Each
COMPARE
Bluetongue Virus 1 No RNA
₹161,000.00INR / Each
COMPARE
Bluetongue Virus 8 No RNA
₹161,000.00INR / Each
COMPARE
Bovine Herpesvirus 1 No DNA
₹161,000.00INR / Each
COMPARE
Bovine Herpesvirus 1 Yes DNA
₹206,000.00INR / Each
COMPARE
Bovine Leukemia Virus No RNA
₹161,000.00INR / Each
Showing 1 - 20 of 23