Cole-Parmer® Biothreat qPCR Reagent Kits
Crucial tools for fast and accurate detection of particularly harmful viruses and bacteria
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
These detection kits utilize the sensitivity and speed of qPCR to get the most accurate data as quickly as possible. Each kit is specifically designed by our bioinformatics team to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. All test kits are validated in house on multiple qPCR platforms to ensure cross platform functionality. Kits come with sufficient controls to reduce the chances of making the wrong call. Kits can also by generated specifically for your test of interest. If the kit is not in our database, we can easily make the kit you wish if sequence data is available. Kits conform to the latest safety directives including European CE requirements.
Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
161,000.00 - ₹352,000.00INR / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Alpha toxin producing Clostridium perfringens No DNA
₹276,000.00INR / Each
COMPARE
Alpha toxin producing Clostridium perfringens Yes DNA
₹352,000.00INR / Each
COMPARE
Bacillus anthracis No DNA
₹161,000.00INR / Each
COMPARE
Bacillus anthracis Yes DNA
₹206,000.00INR / Each
COMPARE
Burkholderia mallei No DNA
₹161,000.00INR / Each
COMPARE
Burkholderia mallei Yes DNA
₹206,000.00INR / Each
COMPARE
Candidatus Mycoplasma haemominutum Yes DNA
₹352,000.00INR / Each
COMPARE
Candidatus Mycoplasma turicensis Yes DNA
₹198,000.00INR / Each
COMPARE
Chlamydophila psittaci No DNA
₹161,000.00INR / Each
COMPARE
Chlamydophila psittaci Yes DNA
₹206,000.00INR / Each
COMPARE
Coxiella burnetii No DNA
₹161,000.00INR / Each
COMPARE
Coxiella burnetii Yes DNA
₹206,000.00INR / Each
COMPARE
Escherichia coli 0157:H7 Yes DNA
₹206,000.00INR / Each
COMPARE
Francisella tularensis No DNA
₹161,000.00INR / Each
COMPARE
Francisella tularensis Yes DNA
₹206,000.00INR / Each
COMPARE
Cryptococcus neoformans No DNA
₹161,000.00INR / Each
COMPARE
Rift Valley Fever Virus No RNA
₹161,000.00INR / Each
COMPARE
Rift Valley Fever Virus Yes RNA
₹206,000.00INR / Each
COMPARE
Vaccinia Virus Yes DNA
₹202,000.00INR / Each
Showing 1- 19 of 19