Cole-Parmer® Ashless Filter Papers
Perfect for gravimetric analysis or sample preparation
  • High-purity, acid-washed filters with high wet strength
Grade 94 filters are fine retention filters for analysis of fine particles such as barium sulfates, lead sulfates, stannic or nickel sulfide, calcium oxalate, and calcium fluoride. Grade 74 filters are well-suited for general filtration such as analysis of clay, copper, bismuth, silicon, and iron. Grade 54 filters are ideal for filtration of coarse or gelatinous precipitate, namely iron, zirconium, or aluminum hydroxides, and cobalt sulfide. Sold 100/pk.
3,800.00 - ₹20,300.00INR / Pkg of 100
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Pore Size (micron) Description
Availability Pricing
COMPARE
240 10 Grade 54
₹20,300.00INR / Pkg of 100
COMPARE
185 1.5 Grade 94
₹14,000.00INR / Pkg of 100
COMPARE
150 10 Grade 54
₹10,400.00INR / Pkg of 100
COMPARE
150 2 Grade 74
₹10,400.00INR / Pkg of 100
COMPARE
90 10 Grade 54
₹6,100.00INR / Pkg of 100
COMPARE
110 1.5 Grade 94
₹7,400.00INR / Pkg of 100
COMPARE
125 10 Grade 54
₹8,300.00INR / Pkg of 100
COMPARE
110 10 Grade 54
₹7,400.00INR / Pkg of 100
COMPARE
55 10 Grade 54
₹3,800.00INR / Pkg of 100
COMPARE
125 1.5 Grade 94
₹8,300.00INR / Pkg of 100
COMPARE
150 1.5 Grade 94
₹10,400.00INR / Pkg of 100
COMPARE
90 2 Grade 74
₹6,100.00INR / Pkg of 100
COMPARE
125 2 Grade 74
₹8,300.00INR / Pkg of 100
COMPARE
90 1.5 Grade 94
₹6,400.00INR / Pkg of 100
COMPARE
110 2 Grade 74
₹7,800.00INR / Pkg of 100
COMPARE
185 2 Grade 74
₹14,000.00INR / Pkg of 100
COMPARE
42.5 1.5 Grade 94
₹4,200.00INR / Pkg of 100
COMPARE
42.5 2 Grade 74
₹4,200.00INR / Pkg of 100
COMPARE
240 2 Grade 74
₹20,300.00INR / Pkg of 100
COMPARE
55 2 Grade 74
₹4,500.00INR / Pkg of 100
Showing 1 - 20 of 22