Cole-Parmer® Aseptic Sampling Assemblies with Nalgene® Media Bottles
One continuous flow path provides aseptic transfer and sampling
  • One-piece molded cap design reduces contamination and product loss
  • Eliminate cleaning validation time
  • Caps feature venting and transferring ports with TPE tubing
  • Bagged and labeled in an ISO Class 7 cleanroom
  • Gamma irradiated to sterility assurance level 10-6
  • USP Class VI and RoHS compliant
  • Lot traceable
These single-use sampling assemblies eliminate setup and after-use cleaning steps required for reusable containers. The aseptic sampling assembly includes sterilized one-piece molded cap with tubing and bottle, providing a closed system to reduce contamination and help protect sample integrity— ideal for bio research, pharmaceutical, and life sciences applications. Cap features two ports with TPE tubing: one for venting and one for transferring.
23,600.00 - ₹46,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Special Features
Availability Pricing
COMPARE
60 Polyethylene Terephthalate Glycol Cap with molded TPE tubing; 8" (20 cm) long feed and 2" (5 cm) long vent
₹23,600.00INR / Each
COMPARE
125 Polyethylene Terephthalate Glycol Cap with molded TPE tubing; 18" (45 cm) long feed and 4" (10 cm) long vent
₹25,400.00INR / Each
COMPARE
250 Polyethylene Terephthalate Glycol Cap with molded TPE tubing; 18" (45 cm) long feed and 4" (10 cm) long vent
₹25,800.00INR / Each
COMPARE
500 Polyethylene Terephthalate Glycol Cap with molded TPE tubing; 18" (45 cm) long feed and 4" (10 cm) long vent
₹28,000.00INR / Each
COMPARE
1000 Polyethylene Terephthalate Glycol Cap with molded TPE tubing; 18" (45 cm) long feed and 4" (10 cm) long vent
₹30,100.00INR / Each
COMPARE
2000 Polycarbonate(PC) Cap with molded TPE tubing; 18" (45 cm) long feed and 4" (10 cm) long vent
₹46,800.00INR / Each