Cole-Parmer® 2-Way, Normally Closed Solenoid Pinch Valves
Precision solenoid pinch valves engineered for fast response and high-cycle operation
  • Fast-response solenoid design supports high-frequency switching and pulse applications
  • Factory-tuned closure ensures a tight, repeatable seal
  • Rated for continuous duty with minimal heat generation
  • Valve rated for up to 20 million cycles; tubing rated for up to 1 million cycles
  • Fluid contacts only the tubing, reducing the risk of contamination
  • Fully unobstructed flow path minimizes pressure drop during operation
  • Compact form factor ideal for space-constrained instruments and assemblies
These 2-way solenoid-operated pinch valves are engineered for applications requiring fast response, repeatable closure, and long service life. Each valve is factory-tuned in a normally closed (NC) configuration to deliver a precise, reliable seal and maintain a fully unobstructed flow path when open.
Designed for continuous switching, these valves support high-frequency operation with minimal heat generation and consistent performance over millions of cycles, making them well suited for pulse dosing, sampling, analytical instrumentation, and integration into compact systems.
18,600.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Pinch Valve Tube Size Power (VDC)
Availability Pricing
COMPARE
15 1/32" ID to 3/32 OD 12
₹18,600.00INR / Each
COMPARE
15 1/16" ID to 1/8" OD 12
₹18,600.00INR / Each
COMPARE
20 1/8" ID to 1/4" OD 12
₹18,600.00INR / Each
COMPARE
15 1/16" ID to 1/8" OD 24
₹18,600.00INR / Each
COMPARE
20 1/8" ID to 1/4" OD 24
₹18,600.00INR / Each