Cole-Parmer® 1-Way Stopcocks with Luer Fittings, Natural Kynar® PVDF
Stopcocks provide accurate, leak-free flow control
  • Handles rotate a full 360°
  • Exhibits exceptional chemical resistance and high purity
  • Meets requirements of ISO 10993-4 and ISO 10993-5 testing
  • Meets USP Class VI, NSF 51, NSF 61, and FDA 21 CFR 177.2510 and RoHS compliant
Add a stopcock to your system for an easy on/off control of your fluid. Stopcocks feature Luer fittings, making it easy to create a quick, leak-free connection. Natural Kynar PVDF stopcocks are animal derivative, DEHP, and BPA free, making them ideal for biomedical research, pharmaceutical, food and beverage applications.
2,200.00 - ₹3,300.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Luer Connection Type (Port 1) Luer Connection Type (Port 2)
Availability Pricing
COMPARE
Female Luer Female Luer
₹2,200.00INR / Pkg of 10
COMPARE
Male Luer Lock Female Luer
₹3,300.00INR / Pkg of 10
COMPARE
Male Lock Luer Male Lock Luer
₹3,300.00INR / Pkg of 10
COMPARE
Male Luer Lock Male Slip Luer
₹3,300.00INR / Pkg of 10
COMPARE
Male Slip Luer Female Luer
₹2,200.00INR / Pkg of 10
COMPARE
Male Slip Luer Male Slip Luer
₹2,200.00INR / Pkg of 10