Clean Room Mats
Maximum performance and quality ensured by consistent adhesive coverage on each layer of matting.
Multi-layer mats are available in 30 or 60 layer versions. Each layer of the mat is fully coated with a particle grabbing adhesive, and an antimicrobial agent that prevents bacterial growth. Mats are constructed of heavy duty 1.8 mil (50 micron) low density polyethylene film. A printed number in the corner of each sheet indicates how many layers remain, and ensures at least one layer remains before replacement. These mats are made in and ISO 9000 factory, which means consistent quality is maintained from box to box.
21,900.00 - ₹66,600.00INR / Pkg of 4
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Width (in) Description
Availability Pricing
Purus PM183634W Cleanroom Mat 18" x 36", 30 Layer, Pk/4 White 6410001 Mfr # PM183634W
COMPARE
36 18 Cleanroom Mat 18" x 36", 30 Layer, Pk/4 White
₹60,900.00INR / Pkg of 4
Purus PM183638W Cleanroom Mat 18" x 36", 30 Layer, Pk/8 White 6410002 Mfr # PM183638W
COMPARE
36 18 Cleanroom Mat 18" x 36", 30 Layer, Pk/8 White
₹45,800.00INR / Pkg of 8
Purus PM243634W Cleanroom Mat 24" x 36", 30 Layer, Pk/4 White 6410011 Mfr # PM243634W
COMPARE
36 24 Cleanroom Mat 24" x 36", 30 Layer, Pk/4 White
₹21,900.00INR / Pkg of 4
Purus PM243638W Cleanroom Mat 24" x 36", 30 Layer, Pk/8 White 6410012 Mfr # PM243638W
COMPARE
36 24 Cleanroom Mat 24" x 36", 30 Layer, Pk/8 White
₹40,800.00INR / Pkg of 8
Purus PM243664W Cleanroom Mat 24" x 36", 60 Layer, Pk/4 White 6410013 Mfr # PM243664W
COMPARE
36 24 Cleanroom Mat 24" x 36", 60 Layer, Pk/4 White
₹40,500.00INR / Pkg of 60
Purus PM264534W Cleanroom Mat 26" x 45", 30 Layer, Pk/4 White 6410021 Mfr # PM264534W
COMPARE
45 26 Cleanroom Mat 26" x 45", 30 Layer, Pk/4 White
₹28,000.00INR / Pkg of 4
Purus PM264538W Cleanroom Mat 26" x 45", 30 Layer, Pk/8 White 6410022 Mfr # PM264538W
COMPARE
45 26 Cleanroom Mat 26" x 45", 30 Layer, Pk/8 White
₹54,500.00INR / Pkg of 8
Purus PM364564W Cleanroom Mat 36" x 45", 60 Layer, Pk/4 White 6410033 Mfr # PM364564W
COMPARE
45 36 Cleanroom Mat 36" x 45", 60 Layer, Pk/4 White
₹66,600.00INR / Pkg of 60
Showing 1- 8 of 8