CELLTREAT Culture Filtration Systems
  • Hydrophilic PES (polyethersulfone) membrane provides fast flow rate
  • Low protein binding—excellent for filtering solutions for cell culture applications
Complete vacuum filtration systems include a polystyrene bottletop filter and solution bottle with high-density polyethylene screw-on cap. Angled hose barb adapter fits 6-, 9-, and 12-mm ID tubing and prevents filter from tipping. Filter systems are supplied sterile (gamma irradiated); packaged individually. Select from 0.22 µm or 0.45 µm pore sizes.
51,700.00 - ₹116,000.00INR / Case of 12
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Membrane Diameter (mm) Pore Size (µm)
Availability Pricing
CELLTREAT Scientific Products 229701 Sterile Filter System with PES membrane, 150 mL, 0.45 µm, 50 mm; 12/cs 0633652 Mfr # 229701
COMPARE
150 50 0.45
₹51,700.00INR / Case of 12
CELLTREAT Scientific Products 229702 Sterile Filter System with PES membrane, 250 mL, 0.45 µm, 50 mm; 12/cs 0633654 Mfr # 229702
COMPARE
250 50 0.45
₹58,200.00INR / Case of 12
CELLTREAT Scientific Products 229703 Sterile Filter System with PES membrane, 500 mL, 0.45 µm, 75 mm; 12/cs 0633656 Mfr # 229703
COMPARE
500 75 0.45
₹81,100.00INR / Case of 12
CELLTREAT Scientific Products 229704 Sterile Filter System with PES membrane, 1000 mL, 0.45 µm, 90 mm; 12/cs 0633658 Mfr # 229704
COMPARE
1000 90 0.45
₹116,000.00INR / Case of 12
CELLTREAT Scientific Products 229705 Sterile Filter System with PES membrane, 150 mL, 0.22 µm, 50 mm; 12/cs 0633660 Mfr # 229705
COMPARE
150 50 0.22
₹52,600.00INR / Case of 12
CELLTREAT Scientific Products 229706 Sterile Filter System with PES membrane, 250 mL, 0.22 µm, 50 mm; 12/cs 0633662 Mfr # 229706
COMPARE
250 50 0.22
₹59,000.00INR / Case of 12
CELLTREAT Scientific Products 229707 Sterile Filter System with PES membrane, 500 mL, 0.22 µm, 75 mm; 12/cs 0633664 Mfr # 229707
COMPARE
500 75 0.22
₹82,000.00INR / Case of 12
CELLTREAT Scientific Products 229708 Sterile Filter System with PES membrane, 1000 mL, 0.22 µm, 90 mm; 12/cs 0633666 Mfr # 229708
COMPARE
1000 90 0.22
₹116,000.00INR / Case of 12
Showing 1- 8 of 8