Burundum™ Grinding Media

Get fast, fine grinding
  • Resist chemical, thermal, and mechanical shock
  • Nonporous, nonconductive, and nonmagnetic
  • Ideal for wet or dry grinding
Media with 1⁄2" (13 mm) dia or smaller pieces are excellent for liquid-based dispersion; media
with 13⁄16" (21 mm) dia pieces are best for grinding dry materials and for general lab use; use
media with 1-1⁄4" (32 mm) dia pieces for grinding and crushing large particles.
91,200.00 - ₹126,000.00INR / Pkg of 940

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Burundum Grinding Media, 13 x 13 mm; 10 lb/Pk
₹91,200.00INR / Pkg of 940
COMPARE
Burundum Grinding Media, 21 x 21 mm; 10 lb/Pk
₹126,000.00INR / Pkg of 180
COMPARE
Burundum Grinding Media, 32 x 32 mm; 10 lb/Pk
₹93,300.00INR / Pkg of 44
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat