Burkle Polypropylene Dipping Beakers
No metal parts to corrode
  • Sample with confidence—convenient strap holds beaker in place
  • Choose from 250-, 600-, or 1000-mL systems
Sample aggressive liquids with these sampling systems made entirely of polypropylene. The rod strap holds the beaker in place while in use, but allows for quick removal for ease of testing afterward.
26,500.00 - ₹28,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
Burkle 5353-0250 Fluid sampling scoop, PP, 100 cm rod, 250 mL WW-06041-96 Mfr # 5353-0250
COMPARE
250
₹26,500.00INR / Each
Burkle 5353-0500 Fluid sampling scoop, PP, 100 cm rod, 600 mL WW-06041-97 Mfr # 5353-0500
COMPARE
600
₹27,400.00INR / Each
Burkle 5353-1000 Fluid sampling scoop, PP, 100 cm rod, 1000 mL WW-06041-98 Mfr # 5353-1000
COMPARE
1000
₹28,100.00INR / Each