Burkle Dry Ice Tablet Maker
Get dry ice tablets in a minute
  • Quick and easy to use
  • No electrical power required
Compact, lightweight dry ice maker allows you to produce a -110°F (-79°C) tablet in approximately one minute. Easily installs on any commercial CO2 gas cylinder with standpipe (thread W 21.8 x 1/14” according to DIN 477-1 No. 6). Ideal for use in laboratories, chemical industry, materials handling facilities, hospitals, universities, schools, and institutes.
The dry ice tablets provide a very low-temperature, low-cost cooling solution for lowering the temperature in cooling baths or cold traps, precooling tissue samples, ensuring safe transport of biological substances, and freezing blood and tissue samples.
126,000.00 - ₹154,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Burkle 9805-0030 Dry Ice Maker, 50 x 30 mm Tablets; 50 g 0604453 Mfr # 9805-0030
COMPARE
Dry Ice Maker, 50 Dia x 30 H mm tablets; 50 g
₹126,000.00INR / Each
Burkle 9805-0200 Dry Ice Maker, 80 x 60 mm tablets; 250 g 0604454 Mfr # 9805-0200
COMPARE
Dry Ice Maker, 80 Dia x 60 H mm tablets; 250 g
₹154,000.00INR / Each