BriskHeat® Silicone Rubber Heating Tapes with SDC Digital Temperature Controller Bundles
Convenient bundle with your choice of heating tape
  • All-inclusive, plug-and-play benchtop temperature controller has everything you need
  • Easy to program and operate
  • Moisture- and chemical-resistant tapes are durable with exceptional flexibility
  • Suitable for electrically conductive surfaces
This bundle starts with your choice of a digital temperature controller that displays its temperature reading in Fahrenheit or Celsius. It has a temperature control range of 32 to 999 °F (0 to 700 °C) with an accuracy rate of +/- 1% full-scale. Includes a type J thermocouple (type K thermocouple is also available; call for details). Dimensions: 5.5" L x 4.25" W x 1.75" H (14.0 x 10.8 x 4.5 cm).
Next, choose a heating tape. They are available in ½" and 1" widths and a variety of lengths and wattages. The tapes are constructed of a silicone rubber outer sheath with a power density of 4.3 watts/in. (0.007 watts/mm). Power leads are 24" (61.0 cm) long with split 2-prong power plug.
111,000.00 - ₹144,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Power (watts) Description
Availability Pricing
BriskHeat SDCJFA-BS0051020L Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 2 ft (13 mm x 0.6 m), 52 Watts, °F Temp Display; 120 VAC 3611974 Mfr # SDCJFA-BS0051020L
COMPARE
450 52 Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 2 ft (13 mm x 0.6 m), 52 Watts, °F Temp Display; 120 VAC
₹111,000.00INR / Each
BriskHeat SDCJFA-BS0051040L Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 4 ft (13 mm x 1.2 m), 104 Watts, °F Temp Display; 120 VAC 3611975 Mfr # SDCJFA-BS0051040L
COMPARE
450 104 Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 4 ft (13 mm x 1.2 m), 104 Watts, °F Temp Display; 120 VAC
₹112,000.00INR / Each
BriskHeat SDCJFA-BS0101100L Silicone Rubber Heating Tape with SDC Temp Controller, 1.0 in x 10 ft (25 mm x 3.1 m), 522 Watts, °F Temp Display; 120 VAC 3611984 Mfr # SDCJFA-BS0101100L
COMPARE
450 522 Silicone Rubber Heating Tape with SDC Temp Controller, 1.0 in x 10 ft (25 mm x 3.1 m), 522 Watts, °F Temp Display; 120 VAC
₹144,000.00INR / Each
BriskHeat SDCJCA-BS0051020L Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 2 ft (13 mm x 0.6 m), 52 Watts, °C Temp Display; 120 VAC 3611990 Mfr # SDCJCA-BS0051020L
COMPARE
450 52 Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 2 ft (13 mm x 0.6 m), 52 Watts, °C Temp Display; 120 VAC
₹111,000.00INR / Each
BriskHeat SDCJCA-BS0051040L Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 4 ft (13 mm x 1.2 m), 104 Watts, °C Temp Display; 120 VAC 3611991 Mfr # SDCJCA-BS0051040L
COMPARE
450 104 Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 4 ft (13 mm x 1.2 m), 104 Watts, °C Temp Display; 120 VAC
₹112,000.00INR / Each
BriskHeat SDCJCA-BS0051060L Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 6 ft (13 mm x 1.8 m), 156 Watts, °C Temp Display; 120 VAC 3611992 Mfr # SDCJCA-BS0051060L
COMPARE
450 156 Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 6 ft (13 mm x 1.8 m), 156 Watts, °C Temp Display; 120 VAC
₹114,000.00INR / Each
BriskHeat SDCJCA-BS0051080L Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in  x 8 ft (13 mm x 2.4 m), 209 Watts, °C Temp Display; 120 VAC 3611993 Mfr # SDCJCA-BS0051080L
COMPARE
450 209 Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 8 ft (13 mm x 2.4 m), 209 Watts, °C Temp Display; 120 VAC
₹119,000.00INR / Each
BriskHeat SDCJCA-BS0051100L Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 10 ft (13 mm x 3.1 m), 261 Watts, °C Temp Display; 120 VAC 3611994 Mfr # SDCJCA-BS0051100L
COMPARE
450 261 Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 10 ft (13 mm x 3.1 m), 261 Watts, °C Temp Display; 120 VAC
₹121,000.00INR / Each
BriskHeat SDCJCA-BS0051120L Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 12 ft (13 mm x 3.7 m), 313 Watts, °C Temp Display; 120 VAC 3611995 Mfr # SDCJCA-BS0051120L
COMPARE
450 313 Silicone Rubber Heating Tape with SDC Temp Controller, 0.5 in x 12 ft (13 mm x 3.7 m), 313 Watts, °C Temp Display; 120 VAC
₹131,000.00INR / Each
BriskHeat SDCJCA-BS0101040L Silicone Rubber Heating Tape with SDC Temp Controller, 1.0 in x 4 ft (25 mm x 1.2 m), 209 Watts, °C Temp Display; 120 VAC 3611997 Mfr # SDCJCA-BS0101040L
COMPARE
450 209 Silicone Rubber Heating Tape with SDC Temp Controller, 1.0 in x 4 ft (25 mm x 1.2 m), 209 Watts, °C Temp Display; 120 VAC
₹123,000.00INR / Each
BriskHeat SDCJCA-BS0101060L Silicone Rubber Heating Tape with SDC Temp Controller, 1.0 in x 6 ft (25 mm x 1.8 m), 313 Watts, °C Temp Display; 120 VAC 3611998 Mfr # SDCJCA-BS0101060L
COMPARE
450 313 Silicone Rubber Heating Tape with SDC Temp Controller, 1.0 in x 6 ft (25 mm x 1.8 m), 313 Watts, °C Temp Display; 120 VAC
₹128,000.00INR / Each
Showing 1- 11 of 11