BriskHeat® Silicone-Rubber Heater Blankets Plastic Surfaces - 120 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 450°F (232°C
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Models for plastic surfaces have a lower 1.25 watts/in2 power density to prevent scorching of the surface
34,000.00 - ₹92,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRP06241 Silicone Heating Blanket, 6x24 Size, 120 Volt, 180 Watt, for plastic surfaces 3600180 Mfr # SRP06241
COMPARE
24 450 120
₹37,200.00INR / Each
BriskHeat SRP12121 Silicone Heating Blanket, 12x12 Size, 120 Volt, 180 Watt, for plastic surfaces 3600184 Mfr # SRP12121
COMPARE
12 450 120
₹34,000.00INR / Each
BriskHeat SRP12361 Silicone Heating Blanket, 12x36 Size, 120 Volt, 540 Watt, for plastic surfaces 3600188 Mfr # SRP12361
COMPARE
36 450 120
₹92,100.00INR / Each