BriskHeat® Silicone-Rubber Heater Blankets Metal Surfaces with Control and Adhesive-120 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 425°F (218°C)
  • Easy-to-apply industrial strength pressure sensitive adhesive backing
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications.
Blanket with Control offers extra convenience and functionality with its included adjustable thermostat control. Models for metal surfaces can be adjusted between 50 to 425°F. No additional control is necessary
The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Heaters for metal surfaces have faster heat-up rates due to the 2.5 watts /in2 power density.
50,700.00 - ₹174,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRL06121PADJB Silicone Heating Blanket with Controller, 50-425F, 6x12 Size, 120 Volt, 180 Watt 3600198 Mfr # SRL06121PADJB
COMPARE
1 425 120
₹50,700.00INR / Each
BriskHeat SRL06241PADJB Silicone Heating Blanket with Controller, 50-425F, 6x24 Size, 120 Volt, 360 Watt 3600201 Mfr # SRL06241PADJB
COMPARE
2 425 120
₹56,300.00INR / Each
BriskHeat SRL06361PADJB Silicone Heating Blanket with Controller, 50-425F, 6x36 Size, 120 Volt, 540 Watt 3600205 Mfr # SRL06361PADJB
COMPARE
3 425 120
₹75,500.00INR / Each
BriskHeat SRL12121PADJB Silicone Heating Blanket with Controller, 50-425F, 12x12 Size, 120 Volt, 360 Watt 3600209 Mfr # SRL12121PADJB
COMPARE
1 425 120
₹58,100.00INR / Each
BriskHeat SRL12241PADJB Silicone Heating Blanket with Controller, 50-425F, 12x24 Size, 120 Volt, 720 Watt 3600213 Mfr # SRL12241PADJB
COMPARE
2 425 120
₹86,500.00INR / Each
BriskHeat SRL12361PADJB Silicone Heating Blanket with Controller, 50-425F, 12x36 Size, 120 Volt, 1080 Watt 3600217 Mfr # SRL12361PADJB
COMPARE
3 425 120
₹126,000.00INR / Each
BriskHeat SRL18181PADJB Silicone Heating Blanket with Controller, 50-425F, 18x18 Size, 120 Volt, 810 Watt 3600221 Mfr # SRL18181PADJB
COMPARE
1.5 425 120
₹99,500.00INR / Each
BriskHeat SRL18361PADJB Silicone Heating Blanket with Controller, 50-425F, 18x36 Size, 120 Volt, 1472 Watt 3600225 Mfr # SRL18361PADJB
COMPARE
3 425 120
₹174,000.00INR / Each
BriskHeat SRL24241PADJB Silicone Heating Blanket with Controller, 50-425F, 24x24 Size, 120 Volt, 1440 Watt 3600229 Mfr # SRL24241PADJB
COMPARE
2 425 120
₹143,000.00INR / Each
Showing 1- 9 of 9