BriskHeat® Silicone-Rubber Heater Blankets Metal Surfaces with Control-120 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 425°F (218°C)
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications.
Blanket with Control offers extra convenience and functionality with its included adjustable thermostat control. Models for metal surfaces can be adjusted between 50 to 425°F. No additional control is necessary
The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Heaters for metal surfaces have faster heat-up rates due to the 2.5 watts /in2 power density.
44,500.00 - ₹156,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRL06121ADJB Silicone Heating Blanket with Controller, for Metals, 6x12" , 120 V, 180 W 3600233 Mfr # SRL06121ADJB
COMPARE
12 425 120
₹44,500.00INR / Each
BriskHeat SRL06241ADJB Silicone Heating Blanket with Controller, for Metals, 6x24" , 120 V, 360 W 3600235 Mfr # SRL06241ADJB
COMPARE
24 425 120
₹54,600.00INR / Each
BriskHeat SRL06361ADJB Silicone Heating Blanket with Controller, for Metals, 6x36" , 120 V, 540 W 3600237 Mfr # SRL06361ADJB
COMPARE
36 425 120
₹68,800.00INR / Each
BriskHeat SRL12121ADJB Silicone Heating Blanket with Controller, for Metals, 12x12" , 120 V, 360 W 3600239 Mfr # SRL12121ADJB
COMPARE
12 425 120
₹54,200.00INR / Each
BriskHeat SRL12241ADJB Silicone Heating Blanket with Controller, for Metals, 12x24" , 120 V, 720 W 3600241 Mfr # SRL12241ADJB
COMPARE
24 425 120
₹74,600.00INR / Each
BriskHeat SRL12361ADJB Silicone Heating Blanket with Controller, for Metals, 12x36" , 120 V, 1080 W 3600243 Mfr # SRL12361ADJB
COMPARE
36 425 120
₹111,000.00INR / Each
BriskHeat SRL18181ADJB Silicone Heating Blanket with Controller, for Metals, 18x18" , 120 V, 810 W 3600245 Mfr # SRL18181ADJB
COMPARE
18 425 120
₹90,100.00INR / Each
BriskHeat SRL18361ADJB Silicone Heating Blanket with Controller, for Metals, 18x36" , 120 V, 1472 W 3600247 Mfr # SRL18361ADJB
COMPARE
36 425 120
₹156,000.00INR / Each
BriskHeat SRL24241ADJB Silicone Heating Blanket with Controller, for Metals, 24x24" , 120 V, 1440 W 3600249 Mfr # SRL24241ADJB
COMPARE
24 425 120
₹126,000.00INR / Each
Showing 1- 9 of 9