BriskHeat Samox® Insulated Heating Tapes with SDC Temp Controller Bundles
No more guesswork with plug-and-play convenience
  • Benchtop On/Off digital temperature controller for simple heating protocols Heavily insulated heating tape protects electrically conductive surfaces Type J thermocouple provides feedback for strict temperature control
All-inclusive bundle features a digital on/off controller with type J thermocouple, heating tape, and power cords—everything you need to get started. Benchtop controller is compact and easy to program. Choose between Fahrenheit and Celsius displays. Heating tapes are insulated with high-temperature Samox. Select heating tape length to optimize coverage while avoiding overlap.
99,400.00 - ₹121,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Power (watts) Width (in)
Availability Pricing
BriskHeat SDCJFA-BWH051020L Samox® Insulated Heating Tape with SDC Temperature Controller, 1400°F Max, 0.50 in x 2 ft (13mm x 0.6m), 156 watts; 120V 3611960 Mfr # SDCJFA-BWH051020L
COMPARE
2 156 1/2
₹99,400.00INR / Each
BriskHeat SDCJFA-BWH051040L Samox® Insulated Heating Tape with SDC Temperature Controller, 1400°F Max, 0.50 in x 4 ft (13mm x 1.2m), 313 watts; 120V 3611961 Mfr # SDCJFA-BWH051040L
COMPARE
4 313 1/2
₹105,000.00INR / Each
BriskHeat SDCJFA-BWH051060L Samox® Insulated Heating Tape with SDC Temperature Controller, 1400°F Max, 0.50 in x 6 ft (13mm x 1.8m), 470 watts; 120V 3611962 Mfr # SDCJFA-BWH051060L
COMPARE
6 470 1/2
₹112,000.00INR / Each
BriskHeat SDCJFA-BWH051080L Samox® Insulated Heating Tape with SDC Temperature Controller, 1400°F Max, 0.50 in x 8 ft (13mm x 2.4m), 627 watts; 120V 3611963 Mfr # SDCJFA-BWH051080L
COMPARE
8 627 1/2
₹117,000.00INR / Each
BriskHeat SDCJFA-BWH101040L Samox® Insulated Heating Tape with SDC Temperature Controller, 1400°F Max, 1.00 in x 4 ft (25mm x 1.2m), 627 watts; 120V 3611965 Mfr # SDCJFA-BWH101040L
COMPARE
4 627 1
₹117,000.00INR / Each
BriskHeat SDCJFA-BWH101060L Samox® Insulated Heating Tape with SDC Temperature Controller, 1400°F Max, 1.00 in x 6 ft (25mm x 1.8m), 940 watts; 120V 3611966 Mfr # SDCJFA-BWH101060L
COMPARE
6 940 1
₹121,000.00INR / Each
BriskHeat SDCJCA-BWH051020L Samox® Insulated Heating Tape with SDC Temperature Controller, 760°C Max, 0.50 in x 2 ft (13mm x 0.6m), 156 watts; 120V 3611967 Mfr # SDCJCA-BWH051020L
COMPARE
2 156 1/2
₹99,400.00INR / Each
BriskHeat SDCJCA-BWH051060L Samox® Insulated Heating Tape with SDC Temperature Controller, 760°C Max, 0.50 in x 6 ft (13mm x 1.8m), 470 watts; 120V 3611969 Mfr # SDCJCA-BWH051060L
COMPARE
6 470 1/2
₹112,000.00INR / Each
BriskHeat SDCJCA-BWH051080L Samox® Insulated Heating Tape with SDC Temperature Controller, 760°C Max, 0.50 in x 8 ft (13mm x 2.4m), 627 watts; 120V 3611970 Mfr # SDCJCA-BWH051080L
COMPARE
8 627 1/2
₹117,000.00INR / Each
BriskHeat SDCJCA-BWH101020L Samox® Insulated Heating Tape with SDC Temperature Controller, 760°C Max, 1.00 in x 2 ft (25mm x 0.6m), 313 watts; 120V 3611971 Mfr # SDCJCA-BWH101020L
COMPARE
2 313 1
₹110,000.00INR / Each
Showing 1- 10 of 10