BriskHeat Heavy-Duty Flexible Drum Heaters with Thermostat
Wrap around a metal or plastic drum to maintain temperature and viscosity
  • Extra-wide 4” heaters provide better surface coverage
  • Built-in thermostat provides adjustable temperature control
  • Resistant to moisture and chemicals
Quickly and efficiently heat-up your metal or plastic drums. These heavy-duty extra-wide silicone rubber band heaters, provides better surface coverage than other traditional drum heaters. The adjustable thermostat control allows the user to set a required temperature. Made of durable vulcanized silicone rubber, fiberglass cloth, and multi-stranded grounded heating elements that provide for a long and reliable service life. The spring closure built-in to the heater allow for expansion up to 3” in order to fit most common drums and pails.
36,800.00 - ₹49,200.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drum Material Drum Size Max Temperature (° F)
Availability Pricing
BriskHeat DHCS10 Flexible 5-Gal Metal Drum Heater with Thermostat; 50-425F, 120V 0310665 Mfr # DHCS10
COMPARE
Metal 15 gallons 425
₹36,800.00INR / Each
BriskHeat DHCS11 Flexible 15-Gal Metal Drum Heater with Thermostat; 50-425F, 120V 0310666 Mfr # DHCS11
COMPARE
Metal 15 gallons 425
₹40,100.00INR / Each
BriskHeat DHCS13 Flexible 30-Gal Metal Drum Heater with Thermostat; 50-425F, 120V 0310667 Mfr # DHCS13
COMPARE
Metal 30 gallons 425
₹45,900.00INR / Each
BriskHeat DHCS15 Flexible 55-Gal Metal Drum Heater with Thermostat; 50-425F, 120V 0310668 Mfr # DHCS15
COMPARE
Metal 55 gallons 425
₹45,700.00INR / Each
BriskHeat DHCS20 Flexible 5-Gal Metal Drum Heater with Thermostat; 50-425F, 240V 0310669 Mfr # DHCS20
COMPARE
Metal 5 gallons 425
₹36,800.00INR / Each
BriskHeat DHCS21 Flexible 15-Gal Metal Drum Heater with Thermostat; 50-425F, 240V 0310670 Mfr # DHCS21
COMPARE
Metal 15 gallons 425
₹40,100.00INR / Each
BriskHeat DHCS23 Flexible 30-Gal Metal Drum Heater with Thermostat; 50-425F, 240V 0310671 Mfr # DHCS23
COMPARE
Metal 30 gallons 425
₹45,900.00INR / Each
BriskHeat DHCS25 Flexible 55-Gal Metal Drum Heater with Thermostat; 50-425F, 240V 0310672 Mfr # DHCS25
COMPARE
Metal 55 gallons 425
₹49,200.00INR / Each
BriskHeat DPCS10 Flexible 5-Gal Poly Drum Heater with Thermostat; 50-160F, 120V 0310673 Mfr # DPCS10
COMPARE
Poly 5 gallons 160
₹36,800.00INR / Each
BriskHeat DPCS11 Flexible 15-Gal Poly Drum Heater with Thermostat; 50-160F, 120V 0310674 Mfr # DPCS11
COMPARE
Poly 15 gallons 160
₹40,100.00INR / Each
BriskHeat DPCS13 Flexible 30-Gal Poly Drum Heater with Thermostat; 50-160F, 120V 0310675 Mfr # DPCS13
COMPARE
Poly 30 gallons 160
₹45,900.00INR / Each
BriskHeat DPCS15 Flexible 55-Gal Poly Drum Heater with Thermostat; 50-160F, 120V 0310676 Mfr # DPCS15
COMPARE
Poly 55 gallons 160
₹49,200.00INR / Each
BriskHeat DPCS20 Flexible 5-Gal Poly Drum Heater with Thermostat; 50-160F, 240V 0310677 Mfr # DPCS20
COMPARE
Poly 5 gallons 160
₹36,800.00INR / Each
BriskHeat DPCS21 Flexible 15-Gal Poly Drum Heater with Thermostat; 50-160F, 240V 0310678 Mfr # DPCS21
COMPARE
Poly 15 gallons 160
₹40,100.00INR / Each
BriskHeat DPCS23 Flexible 30-Gal Poly Drum Heater with Thermostat; 50-160F, 240V 0310679 Mfr # DPCS23
COMPARE
Poly 30 gallons 160
₹45,900.00INR / Each
BriskHeat DPCS25 Flexible 55-Gal Poly Drum Heater with Thermostat; 50-160F, 240V 0310682 Mfr # DPCS25
COMPARE
Poly 55 gallons 160
₹49,200.00INR / Each
Showing 1- 16 of 16