BriskHeat® Full Coverage Insulated Drum Heaters
Provides safe, uniform heat to 55-gallon metal or plastic drums
  • Built-in adjustable thermoswitch prevents temperature runaway Lightweight flexible design makes installation quick and easy Order Insulation Cover (09300-70) separately
These insulated drum heaters provide a safe and efficient method of temperature and viscosity control for liquids in 55-gallon drums. Simply wrap insulated drum heater around a 55-gallon drum, adjust the built-in thermostat to desired temperature setting, and uniform heat is evenly distributed around the entire drum. The grounded heating element provides a high-watt density, ensuring rapid heating times — one inch thick fiberglass insulation prevents heat loss. Prevent further heat loss by ordering the drum insulation top cover (optional) below.
Drum heaters feature a hook-and-loop seal for easy installation and removal and models are available for both metal and plastic 55-gallon drums. Heaters come with standard three-prong plug (240 VAC models do not come with plugs) and are intended for indoor use in non-hazardous areas only.
310,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drum Material Drum Size Max Temperature (° F)
Availability Pricing
BriskHeat FGDHC55120 55 gallon, Full Coverage, Drum Heater - 120V 0930050 Mfr # FGDHC55120D
COMPARE
Metal 55 gallons 450
₹310,000.00INR / Each
BriskHeat FGDHC55240 55 Gallon, Full Coverage, Metal Drum Heater, Dual Zone; 240 V 0930055 Mfr # FGDHC55240D
COMPARE
Metal 55 gallons 450
₹310,000.00INR / Each
BriskHeat FGPDHC55120 55gallon, Full Coverage, Drum Heater - 120 V 0930060 Mfr # FGPDHC55120
COMPARE
Plastic 55 gallons 160
₹310,000.00INR / Each
BriskHeat FGPDHC55240D 55gallon, Full Coverage, Plastic Drum Heater, Single Zone- 240 V 0930065 Mfr # FGPDHC55240D
COMPARE
Plastic 55 gallons 160
₹310,000.00INR / Each