BriskHeat® Fibrox High-Temperature Heating Tapes with Leads on Opposite Ends-240 VAC
Flexible, multipurpose, direct-contact heaters are ideal for use on conductive surfaces
  • Maximum exposure temperature of 900°F (482°C)
  • Choose from power leads on same end or on opposite ends
  • Heavy-duty insulated design
  • Nongrounded
These high-temperature heating tapes are made of resistance wires that are insulated with braided fiberglass knitted into a flat tape and covered with heavy fiberglass fabric.
The 240 VAC tapes with an opposite-end lead configuration are supplied bare leads.
Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.
11,800.00 - ₹46,200.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (watts)
Availability Pricing
BriskHeat TBIH 052-020 Fibrox Insulated Heating Tape, Leads on Opposite Ends, 2 ft L x 1/2"W; 240 VAC 0310521 Mfr # TBIH 052-020
COMPARE
2 900 104
₹11,800.00INR / Each
BriskHeat TBIH 052-040 Fibrox Insulated Heating Tape, Leads on Opposite Ends, 4 ft L x 1/2"W; 240 VAC 0310541 Mfr # TBIH 052-040
COMPARE
4 900 208
₹14,600.00INR / Each
BriskHeat TBIH 102-100 Fibrox Insulated Heating Tape, Leads on Opposite Ends, 10 ft L x 1"W; 240 VAC 0310610 Mfr # TBIH 102-100
COMPARE
10 900 1045
₹46,200.00INR / Each
BriskHeat TBIH-102-040 Fibrox Insulated Heating Tape, Leads on Opposite Ends, 4 ft L x 1"W; 240 VAC 0310641 Mfr # TBIH-102-040
COMPARE
4 900 420
₹19,700.00INR / Each