BriskHeat Fiberglass Insulated Heating Tapes with SDC Temp Controller Bundles
No more guesswork with plug-and-play convenience
  • Benchtop On/Off digital temperature controller for simple heating protocols Heavily insulated heating tape protects electrically conductive surfaces Type J thermocouple provides feedback for strict temperature control
All-inclusive bundle features a digital on/off controller with type J thermocouple, heating tape, and power cords—everything you need to get started. Benchtop controller is compact and easy to program. Choose between Fahrenheit and Celsius displays. Heating tapes are insulated with standard fiberglass. Select heating tape length to optimize coverage while avoiding overlap.
103,000.00 - ₹126,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Power (watts) Width (in)
Availability Pricing
BriskHeat SDCJFA-BIH051040L F Fiberglass Insulated Heating Tape with SDC Temperature Controller, 900°F Max, 0.50 in x 4 ft (13mm x 1.2m), 210 watts; 120V 3611920 Mfr # SDCJFA-BIH051040L F
COMPARE
4 210 1/2
₹106,000.00INR / Each
BriskHeat SDCJFA-BIH051060L F Fiberglass Insulated Heating Tape with SDC Temperature Controller, 900°F Max, 0.50 in x 6 ft (13mm x 1.8m), 310 watts; 120V 3611921 Mfr # SDCJFA-BIH051060L F
COMPARE
6 310 1/2
₹112,000.00INR / Each
BriskHeat SDCJFA-BIH051080L F Fiberglass Insulated Heating Tape with SDC Temperature Controller, 900°F Max, 0.50 in x 8 ft (13mm x 2.4m), 420 watts; 120V 3611922 Mfr # SDCJFA-BIH051080L F
COMPARE
8 420 1/2
₹112,000.00INR / Each
BriskHeat SDCJFA-BIH101040L F Fiberglass Insulated Heating Tape with SDC Temperature Controller, 900°F Max, 1.00 in x 4 ft (25mm x 1.2m), 420 watts; 120V 3611927 Mfr # SDCJFA-BIH101040L F
COMPARE
4 420 1
₹110,000.00INR / Each
BriskHeat SDCJFA-BIH101060L F Fiberglass Insulated Heating Tape with SDC Temperature Controller, 900°F Max, 1.00 in x 6 ft (25mm x 1.8m), 620 watts; 120V 3611928 Mfr # SDCJFA-BIH101060L F
COMPARE
6 620 1
₹117,000.00INR / Each
BriskHeat SDCJCA-BIH051040L C Fiberglass Insulated Heating Tape with SDC Temperature Controller, 482°C Max, 0.50 in x 4 ft (13mm x 1.2m), 210 watts; 120V 3611949 Mfr # SDCJCA-BIH051040L C
COMPARE
4 210 1/2
₹106,000.00INR / Each
BriskHeat SDCJCA-BIH051060L C Fiberglass Insulated Heating Tape with SDC Temperature Controller, 482°C Max, 0.50 in x 6 ft (13mm x 1.8m), 310 watts; 120V 3611950 Mfr # SDCJCA-BIH051060L C
COMPARE
6 310 1/2
₹109,000.00INR / Each
BriskHeat SDCJCA-BIH051120L C Fiberglass Insulated Heating Tape with SDC Temperature Controller, 482°C Max, 0.50 in x 12 ft (13mm x 3.7m), 620 watts; 120V 3611953 Mfr # SDCJCA-BIH051120L C
COMPARE
12 620 1/2
₹126,000.00INR / Each
BriskHeat SDCJCA-BIH101010L C Fiberglass Insulated Heating Tape with SDC Temperature Controller, 482°C Max, 1.00 in x 1 ft (25mm x 0.3m), 105 watts; 120V 3611954 Mfr # SDCJCA-BIH101010L C
COMPARE
1 105 1
₹103,000.00INR / Each
BriskHeat SDCJCA-BIH101040L C Fiberglass Insulated Heating Tape with SDC Temperature Controller, 482°C Max, 1.00 in x 4 ft (25mm x 1.2m), 420 watts; 120V 3611956 Mfr # SDCJCA-BIH101040L C
COMPARE
4 420 1
₹110,000.00INR / Each
Showing 1- 10 of 10