BriskHeat® Dual-Element Heating Tapes with Benchtop Temperature Controller - Celsius - 120 VAC
Convenient heating system that includes on/off temperature controller, flexible heating tape, and temperature sensor
  • Celsius controller temperature range 0 to 600°C
  • Factory-installed 5-foot J-Type thermocouple sensor
The flexible dual-element heating tape wraps around and heats a wide array of tubing, vessels, and apparatuses. The extremely flexible and high-temperature heating tape is made of two heating elements that are knitted together in series and then wound together with high-temperature Samox fiberglass.
The included digital on/off temperature controller provides basic control and connects to the heating tape without any additional wiring. The large display is easy to read and simple to program. Accuracy is +/- 1% FS. Hysteresis is adjustable between 1 and 99 degrees. In the case of probe failure, heating can be selected on, off, or periodical running. It is benchtop-friendly with its compact 5.5” x 4.25” x 1.75” profile. The controller has a 10amp fused output and can be used with other heating tapes, beaker heaters, and mantle heaters as long as they are equipped with standard U.S. plugs.
Approvals and Compliances: Controller: CE
Heating Tape: CE, RoHS, REACH
Warning: Heating tapes should not be overlapped.
89,300.00 - ₹108,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° C) Power (VAC)
Availability Pricing
BriskHeat CPSDCJCA-BWH051020LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1/2" x 24" , 120 V 3600348 Mfr # CPSDCJCA-BWH051020LD
COMPARE
24 600 120
₹89,300.00INR / Each
BriskHeat CPSDCJCA-BWH051040LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1/2" x 48" , 120 V 3600350 Mfr # CPSDCJCA-BWH051040LD
COMPARE
48 600 120
₹92,300.00INR / Each
BriskHeat CPSDCJCA-BWH051060LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1/2" x 72" , 120 V 3600352 Mfr # CPSDCJCA-BWH051060LD
COMPARE
72 600 120
₹96,300.00INR / Each
BriskHeat CPSDCJCA-BWH051080LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1/2" x 96" , 120 V 3600354 Mfr # CPSDCJCA-BWH051080LD
COMPARE
96 600 120
₹101,000.00INR / Each
BriskHeat CPSDCJCA-BWH051100LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1/2" x 120" , 120 V 3600356 Mfr # CPSDCJCA-BWH051100LD
COMPARE
120 600 120
₹108,000.00INR / Each
BriskHeat CPSDCJCA-BWH101020LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1" x 24" , 120 V 3600358 Mfr # CPSDCJCA-BWH101020LD
COMPARE
24 600 120
₹93,500.00INR / Each
BriskHeat CPSDCJCA-BWH101040LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1" x 48" , 120 V 3600360 Mfr # CPSDCJCA-BWH101040LD
COMPARE
48 600 120
₹94,600.00INR / Each
BriskHeat CPSDCJCA-BWH101060LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1" x 72" , 120 V 3600362 Mfr # CPSDCJCA-BWH101060LD
COMPARE
72 600 120
₹96,700.00INR / Each
Showing 1- 8 of 8