BriskHeat® Dual-Element Heating Tapes - 240 VAC
Greater flexibility than other high-temperature heating tapes
  • Heat to 1400°>F (760°>C) ideal for high-temperature use on conductive surfaces
Dual-element design improves the heater’s flexibility over standard high-temperature
tapes. Tapes are made of two heating elements that are knitted together in series and then
wound together with high-temperature Samox® fiberglass. The 1240 VAC tapes are supplied with bare leads.
Warning: Heating tapes should not be overlapped and require the use of a temperature or
power controller sold separately.
What’s included: reusable tie straps
19,700.00 - ₹44,100.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat TBIH052-040LD Dual Element, Cloth Insulated Heat Tape, 4' x 1/2" , 312 Watts, 240 V 3605015 Mfr # TBIH052-040LD
COMPARE
4 1400 240
₹19,700.00INR / Each
BriskHeat TBIH052-060LD Dual Element, Cloth Insulated Heat Tape, 6' x 1/2" , 468 Watts, 240 V 3605025 Mfr # TBIH052-060LD
COMPARE
6 1400 240
₹24,600.00INR / Each
BriskHeat TBIH052-080LD Dual Element, Cloth Insulated Heat Tape, 8' x 1/2" , 624 Watts, 240 V 3605035 Mfr # TBIH052-080LD
COMPARE
8 1400 240
₹28,200.00INR / Each
BriskHeat TBIH052-100LD Dual Element, Cloth Insulated Heat Tape, 10' x 1/2" , 780 Watts, 240 V 3605045 Mfr # TBIH052-100LD
COMPARE
10 1400 240
₹31,300.00INR / Each
BriskHeat TBIH052-120LD Dual Element, Cloth Insulated Heat Tape, 12' x 1/2" , 936 Watts, 240 V 3605055 Mfr # TBIH052-120LD
COMPARE
12 1400 240
₹34,400.00INR / Each
BriskHeat TBIH102-020LD Dual Element, Cloth Insulated Heat Tape, 2' x 1" , 312 Watts, 240 V 3605065 Mfr # TBIH102-020LD
COMPARE
2 1400 240
₹22,200.00INR / Each
BriskHeat TBIH102-040LD Dual Element, Cloth Insulated Heat Tape, 4' x 1" , 624 Watts, 240 V 3605075 Mfr # TBIH102-040LD
COMPARE
4 1400 240
₹24,900.00INR / Each
BriskHeat TBIH102-060LD Dual Element, Cloth Insulated Heat Tape, 6' x 1" , 936 Watts, 240 V 3605085 Mfr # TBIH102-060LD
COMPARE
6 1400 240
₹44,100.00INR / Each
BriskHeat TBIH102-080LD Dual Element, Cloth Insulated Heat Tape, 8' x 1" , 1248 Watts, 240 V 3605095 Mfr # TBIH102-080LD
COMPARE
8 1400 240
₹34,800.00INR / Each
Showing 1- 9 of 9