BrandTech® Seripettor® Pro Dispensers
Wider range of liquids than standard seripettor model
  • Replaceable dispensing cartridge keeps service costs down
  • Opaque upper sleeve protects light-sensitive reagents
These seripettor® pro dispensers offer the rugged versatility and low lifetime cost of the regular seripettor and handle a wider range of liquids. Ideal for customers who do not require all the features of the Dispensette® dispensers. Seripettor pro dispensers are threaded to fit directly onto 45-mm screw-neck bottles; adapters for S40, 33- and 38-mm bottles are included. Order bottles separately.
113,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Division (mL) Max Dispense Volume (mL) Min Dispense Volume (mL)
Availability Pricing
BrandTech 4720420 Seripettor® Pro Dispenser, 0.2 to 2 mL 0787827 Mfr # 4720420
COMPARE
0.2 2 0.2
₹113,000.00INR / Each