BrandTech® Seripettor® Economy Dispensers
An economical choice for dispensing of many liquids in biological labs
  • Spring-loaded piston for one handed dispensing
  • Replaceable piston/cylinder cartridge to renew dispenser
  • Suitable for sodium azide solutions
The seripettor® is an economical dispenser suitable for many needs of the biology laboratory. An excellent choice for bases, buffers, and other aqueous solutions as well as dilute, non-oxidizing acids, and polar solvents. Wetted materials include PP, HDPE, and EPDM rubber. Not recommended for concentrated acids, non-polar solvents, or scintillation fluid. Accessories are available for dispensing of culture media or agar.
44,800.00 - ₹57,500.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Division (mL) Max Dispense Volume (mL) Min Dispense Volume (mL)
Availability Pricing
BrandTech Seripettor Economy Dispenser, 0.2 to 2 mL 0787828 Mfr # 4720120
COMPARE
0.04 2 0.2
₹44,800.00INR / Each
BrandTech Seripettor Economy Dispenser, 1.0 to 10 mL 0787830 Mfr # 4720140
COMPARE
0.2 10 1
₹44,800.00INR / Each
BrandTech Seripettor Economy Dispenser, 2.5 to 25 mL 0787832 Mfr # 4720150
COMPARE
0.5 25 2.5
₹57,500.00INR / Each