BrandTech® PD-Tip™ II Pipette Tips
Fits all repeating pipettes using standard tips
  • High quality virgin plastic construction
  • Encoded for automatic recognition
  • DNA, RNAse, endotoxin, and ATP free
  • Available sterile and non-sterile
PD-Tip™ Precision Dispenser tips are constructed of high-quality virgin high-density polyethylene and polypropylene (0.1 mL tip is polypropylene/LCP). An extended piston minimizes dead volume and maximizes fluid utilization. The precision seal between the piston and cylinder ensures smooth operation, increased accuracy, and increased precision while pipetting. Sterile tips are individually wrapped and certified free of DNA, RNAse, endotoxins, and ATP. 25 and 50 mL tip packs include one autoclavable, polypropylene adapter.
Size automatically recognized by BRAND® HandyStep® touch and electronic, Rainin® AutoRep E and Gilson® Repetman. Compatible with pipettes from BRAND®, Rainin, Gilson (Repetman), Bel-Art and others.
11,700.00 - ₹48,200.00INR / Pkg of 140
23 variations of this product are available.
See All Product Options Below
23 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sterile Tip Volume (mL) Wetted Matterial
Availability Pricing
BrandTech PD-Tip II Pipette Tips, Set, Non-Sterile, 0.5, 1, 1.25, 2.5, 5, 10, 12.5 mL; 20/PK 2480606 Mfr # 705720
COMPARE
No 12.5 Polyethylene, polypropylene, and liquid crystal polymer
₹47,000.00INR / Pkg of 140
BrandTech PD-Tip II Pipette Tips, Non-Sterile, 0.5 mL; 100/PK 2480607 Mfr # 705702
COMPARE
No 0.5 Polyethylene and polypropylene
₹35,300.00INR / Pkg of 100
BrandTech PD-Tip II Pipette Tips, Non-Sterile, 1.25 mL; 100/PK 2480608 Mfr # 705706
COMPARE
No 1.25 Polyethylene and polypropylene
₹35,300.00INR / Pkg of 100
BrandTech PD-Tip II Pipette Tips, Non-Sterile, 2.5 mL; 100/PK 2480609 Mfr # 705708
COMPARE
No 2.5 Polyethylene and polypropylene
₹35,700.00INR / Pkg of 100
BrandTech PD-Tip II Pipette Tips, Non-Sterile, 5 mL; 100/PK 2480610 Mfr # 705710
COMPARE
No 5 Polyethylene and polypropylene
₹35,700.00INR / Pkg of 100
BrandTech PD-Tip II Pipette Tips, Non-Sterile, 12.5 mL; 100/PK 2480611 Mfr # 705714
COMPARE
No 12.5 Polyethylene and polypropylene
₹35,300.00INR / Pkg of 100
BrandTech PD-Tip II Pipette Tips, Non-Sterile, 25 mL; 50/PK 2480612 Mfr # 705716
COMPARE
No 25 Polyethylene and polypropylene
₹23,800.00INR / Pkg of 50
BrandTech PD-Tip II Pipette Tips, Non-Sterile, 50 mL; 25/PK 2480613 Mfr # 705718
COMPARE
No 50 Polyethylene and polypropylene
₹11,700.00INR / Pkg of 25
BrandTech PD-Tip II Pipette Tips, Non-Sterile Adapter for 25 & 50 mL Tips; 10/PK 2480614 Mfr # 702398
COMPARE
No 50 Polypropylene
₹28,600.00INR / Pkg of 10
BrandTech PD-Tip II Pipette Tips, Non-Sterile, 0.1 mL; 100/PK 2480615 Mfr # 705700
COMPARE
No 0.1 Polypropylene and liquid crystal polymer
₹35,300.00INR / Pkg of 100
BrandTech PD-Tip II Pipette Tips, Non-Sterile, 1 mL; 100/PK 2480616 Mfr # 705704
COMPARE
No 1 Polyethylene and polypropylene
₹35,300.00INR / Pkg of 100
BrandTech PD-Tip II Pipette Tips, Non-Sterile, 10 mL; 100/PK 2480617 Mfr # 705712
COMPARE
No 10 Polyethylene and polypropylene
₹35,700.00INR / Pkg of 100
BrandTech PD-Tip II BIO-CERT® Pipette Tips, Sterile, 0.1 mL; 100/PK 2480618 Mfr # 705730
COMPARE
Yes 0.1 Polypropylene and liquid crystal polymer
₹48,200.00INR / Pkg of 100
BrandTech PD-Tip II BIO-CERT® Pipette Tips, Sterile, 0.5 mL; 100/PK 2480619 Mfr # 705732
COMPARE
Yes 0.5 Polyethylene and polypropylene
₹48,200.00INR / Pkg of 100
BrandTech PD-Tip II BIO-CERT® Pipette Tips, Sterile, 1 mL; 100/PK 2480620 Mfr # 705734
COMPARE
Yes 1 Polyethylene and polypropylene
₹48,200.00INR / Pkg of 100
BrandTech PD-Tip II BIO-CERT® Pipette Tips, Sterile, 1.25 mL; 100/PK 2480621 Mfr # 705736
COMPARE
Yes 1.25 Polyethylene and polypropylene
₹48,200.00INR / Pkg of 100
BrandTech PD-Tip II BIO-CERT® Pipette Tips, Sterile, 2.5 mL; 100/PK 2480622 Mfr # 705738
COMPARE
Yes 2.5 Polyethylene and polypropylene
₹48,200.00INR / Pkg of 100
BrandTech PD-Tip II BIO-CERT® Pipette Tips, Sterile, 5 mL; 100/PK 2480623 Mfr # 705740
COMPARE
Yes 5 Polyethylene and polypropylene
₹48,200.00INR / Pkg of 100
BrandTech PD-Tip II BIO-CERT® Pipette Tips, Sterile, 10 mL; 100/PK 2480624 Mfr # 705742
COMPARE
Yes 10 Polyethylene and polypropylene
₹48,200.00INR / Pkg of 100
BrandTech PD-Tip II BIO-CERT® Pipette Tips, Sterile, 12.5 mL; 100/PK 2480625 Mfr # 705744
COMPARE
Yes 12.5 Polyethylene and polypropylene
₹48,200.00INR / Pkg of 100
Showing 1 - 20 of 23