BrandTech® Disposable Polystyrene Spectrophotometry Cuvettes
Mold-cavity matched for consistency
  • Superior results in the visible spectrum (=340 nm)
  • Expanded polystyrene packaging to preserve optical quality
Each and every BRAND cuvette benefits from their over quarter century of experience in precision cuvette molding to the most exacting standards for reproducible results. Cuvettes have 10 mm optical pathlength, and are available in macro (2.5 mL =- 4.5 mL) and semi-micro (1.5 mL - 3.0 mL) sizes.
6,600.00 - ₹30,200.00INR / Pkg of 500
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Capacity (mL) Pathlength (mm)
Availability Pricing
BrandTech Macro Polystyrene (PS) Cuvettes, 500/PK 0634351 Mfr # 759070D
COMPARE
Polystyrene 4.5 10
₹27,700.00INR / Pkg of 500
BrandTech Macro Polystyrene (PS) Cuvettes, 100/PK 0634352 Mfr # 759071D
COMPARE
Polystyrene 4.5 10
₹6,600.00INR / Pkg of 100
BrandTech Semi-Micro Polystyrene (PS) Cuvettes, 500/PK 0634353 Mfr # 759075D
COMPARE
Polystyrene 3 10
₹30,200.00INR / Pkg of 500
BrandTech Semi-Micro Polystyrene (PS) Cuvettes, 100/PK 0634354 Mfr # 759076D
COMPARE
Polystyrene 3 10
₹6,900.00INR / Pkg of 100