Boekel Wax Dispensers
Prevent wax blockage with heated tap
  • Insulated inner reservoir provides excellent thermal transfer and temperature control
  • Inner screen prevents large particulates from clogging the dispenser valve
  • Enhanced safety with high temperature cut-off
The Boekel paraffin wax dispensers provide fast heating for on-demand molten wax, ideal for your histology and pathology applications. The heated tap prevents blockage providing a smooth even flow of wax every time. An internal filter screen prevents large particles from clogging the tap, ensuring wax flows quickly and consistently.
663,000.00 - ₹1,913,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (L) Max Temperature (° C) Power (VAC)
Availability Pricing
Boekel Large Wax Dispenser, 28 L; 115 VAC, 50/60 Hz 0110015 Mfr # 1456XL
COMPARE
28 65 115
₹1,913,000.00INR / Each
Boekel Large Wax Dispenser, 28 L; 230 VAC, 50/60 Hz 0110016 Mfr # 1456XL-2
COMPARE
28 65 230
₹1,913,000.00INR / Each
Boekel Wax Dispenser, 5 L; 115 VAC, 50/60 Hz 0110017 Mfr # 145600
COMPARE
5 75 115
₹663,000.00INR / Each
Boekel Wax Dispenser, 5 L; 230 VAC, 50/60 Hz 0110018 Mfr # 145600-2
COMPARE
5 75 230
₹689,000.00INR / Each