Boekel Electronic Desiccator with Flexboard Shelves
Don’t keep replacing desiccant
  • Automatic regeneration of desiccant every five hours
  • Sturdy, stainless steel and glass construction
  • Monitor humidity with included hygrometer
No wasted space—desiccant housed in side mount box for more sample space.
1,282,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Boekel Scientific 134041-2 Electronic Desiccator with Flexboard Shelves, 230 VAC 0890125 Mfr # 134041-2
COMPARE
230
₹1,282,000.00INR / Each