Binder Solid.Line Gravity Convection Ovens
Count on accurate temperature controls
  • Controller with timer function simplifies customized use
  • Adjustable exhaust air flap accommodates your environment
Enjoy the reliability required in research, quality assurance and other key applications. The APT.line™ preheating chamber technology ensures precise temperatures, while the independently adjustable ISO Class 2 safety thermostat with visual alarm protects against overheating. Allowing for stacking, each oven offers ergonomic door opening and easy handling.
313,000.00 - ₹456,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC) Rise Time
Availability Pricing
Binder ED-S 56 Solid.Line Gravity Convection Oven, 2.2 cu ft; 120V WW-05012-68 Mfr # 9090-0015
COMPARE
2.2 120 60 mins to 150°C
₹313,000.00INR / Each
Binder ED-S 56 Solid.Line Gravity Convection Oven, 2.2 cu ft; 230V WW-05012-69 Mfr # 9090-0014
COMPARE
2.2 230 60 mins to 150°C
₹365,000.00INR / Each
Binder ED-S 115 Solid.Line Gravity Convection Oven, 4.2 cu ft; 230V WW-05012-72 Mfr # 9090-0020
COMPARE
4.2 230 55 mins to 150°C
₹456,000.00INR / Each