Bi-Therm Superior Grade Bimetal Thermometers

No more inaccurate temperature readings
  • Calibrate thermometer yourself for greater accuracy
  • Three-point calibration assures ±2° accuracy across entire range
  • Stainless steel 8" stem features a dimple indicating how deep to insert for the most accurate result
4,300.00 - ₹5,400.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Dial Size (in) Max Temperature (° F) Min Temperature (° F)
Availability Pricing
Taylor 8215N Dial Bi-Metal Test Thermometer, 1 3/4", 8" Stem, 0 to 220°F 9000103 Mfr # 8215N
COMPARE
1.75 220 0
₹4,300.00INR / Each
Taylor 6215J 2" Dial Bi-metal Test Thermometer, 8" Stem, 0/220F, 1% Accy 9000106 Mfr # 6215J
COMPARE
2 220 0
₹5,400.00INR / Each
Taylor 6220J 2" Dial Bi-metal Test Thermometer, 8" Stem, 50F/550F, 1% Accy 9000107 Mfr # 6220J
COMPARE
2 500 50
₹5,400.00INR / Each
Taylor 6235J 2" Dial Bi-metal Test Thermometer, 8" Stem, -10C/110C, 1% Accy 9000108 Mfr # 6235J
COMPARE
2 230 14
₹4,600.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat