BEVCO Vinyl Cleanroom Stools
Durable 16'' diameter seat provides comfortable seating and easy under-counter storage
Stools are constructed of a sturdy, five-legged base in polished aluminum or chrome tubular steel. Easy-to-use pneumatic seat adjusts in height for variance in worker size. Adjustable foot ring measures 18'' in diameter — set it for the most ergonomic sitting position. All models are black and include mushroom glides for stability. Order optional casters separately.
Vinyl Stools feature dependable vinyl over soft foam padding.
Compliance: ISO Class 5 & CAL 117
140,000.00 - ₹155,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Seat height (in) Description
Availability Pricing
Bevco 3050C1V+ACF-18 BLACK Tall Height Cleanroom Stool, Vinyl, 15.5" to 20.5" height, aluminum base 8652001 Mfr # 3050C2-V-BLK-ACF-18
COMPARE
Vinyl 15.5 to 20.5 Versa Tall Height ISO 5 Cleanroom Black Vinyl Backless Stool; Polished Aluminum Base w/Adjustable Footring
₹140,000.00INR / Each
Bevco 3610C1V-BLACK Mid Height Cleanroom stool, vinyl, 25" to 30", chrome tubular steel base 8652002 Mfr # 3610C2-V-BLK
COMPARE
Vinyl 24 to 29 Versa Mid Height ISO 5 Cleanroom Black Vinyl Backless Stool; Chrome Tubular Steel Base w/Welded Footring
₹155,000.00INR / Each
Bevco 3350C1V+ACF-18 BLACK Mid Height Cleanroom Stool, Vinyl, 18.5" to 26" height, aluminum base 8652003 Mfr # 3350C2-V-BLK-ACF-18
COMPARE
Vinyl 18.5 to 26 Versa Mid Height ISO 5 Cleanroom Black Vinyl Backless Stool; Polished Aluminum Base w/Adjustable Footring
₹144,000.00INR / Each