BEVCO ESD Vinyl Cleanroom Stools
Durable 16'' diameter seat meets ESD/ISO needs, provides comfortable seating, and easy under-counter storage
  • 300 lb weight capacity
Stools are constructed of a sturdy, five-legged base in polished aluminum or chrome tubular steel. Easy-to-use pneumatic seat adjusts in height for variance in worker size. Adjustable foot ring measures 18'' in diameter — set it for the most ergonomic sitting position. All models are black and include mushroom glides for stability. Order optional casters separately below.
ESD (Electrostatic Discharge) Vinyl Stools also include a brass drag chain and ESD mushroom glides.
ESD/ISO 5 Cleanroom Series models are designed to meet specific particulate goals of ISO 14644-1 and the ESD requirements of ANSI/ESD STM12.1-2013 delivering a combination ESD/Cleanroom seating solution for highly technical environments. The vinyl seat complies with CAL 117.
170,000.00 - ₹183,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Material Seat height (in) Description
Availability Pricing
Bevco 3050E1V+ACF-18E BLACK Cleanroom Stool, ESD Vinyl, 15.5" to 20.5" height, aluminum base 8652011 Mfr # 3050E2-V-BLK-ACF-18E
COMPARE
ESD Vinyl 15.5 to 20.5 Versa Desk Height ESD/ISO 5 Cleanroom Black Vinyl Backless Stool; Polished Aluminum Base w/Adjustable Footring
₹170,000.00INR / Each
Bevco 3610E1V-BLACK Cleanroom stool, ESD vinyl, 24" to 29" height, 22" diameter chrome tubular steel 8652012 Mfr # 3610E2-V-BLK
COMPARE
ESD Vinyl 24 to 29 Versa Tall Height ESD/ISO 5 Cleanroom Black Vinyl Backless Stool; Chrome Tubular Steel Base w/Welded Footring
₹183,000.00INR / Each
Bevco 3350E1V+ACF-18E BLACK Cleanroom Stool, ESD Vinyl, 18.5" to 26" height, aluminum base 8652013 Mfr # 3350E2-V-BLK-ACF-18E
COMPARE
ESD Vinyl 18.5 to 26 Versa Mid Height ESD/ISO 5 Cleanroom Black Vinyl Backless Stool; Polished Aluminum Base wAdjustable Footring
₹174,000.00INR / Each
Bevco 3550E1V+ACF-18E-BLACK Cleanroom stool, ESD vinyl, 21.5" to 31.5" height, chrome tubular steel base 8652015 Mfr # 3550E2-V-BLK-ACF-18E
COMPARE
ESD Vinyl 21.5 to 31.5 Versa Tall Height ESD/ISO 5 Cleanroom Black Vinyl Backless Stool; Polished Aluminum Base wAdjustable Footring
₹179,000.00INR / Each