Bel-Art Poxygrid Benchtop Biohazard Bag Holders
Comply with OSHA standards for biohazardous material handling
  • Epoxy-coated steel wire stand is durable and easy to clean
  • All components are autoclavable
These orange colored epoxy-coated steel wire stands can securely hold a high-density 8 1/2" x 11" (22 x 28 cm) polyethylene bag in an upright open position. Ideal for collecting pipette tips, microcentrifuge tubes, and other hazardous materials throughout the day. The stands are reusable, can be cleaned by hand or in automated dish washer, and autoclavable at 250°F (121°C).
7,500.00 - ₹12,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Width (in)
Availability Pricing
Bel-Art Poxygrid Benchtop Biohazard Bag Holder Kit WW-06496-01 Mfr # F13193-0500
COMPARE
11 Epoxy-coated steel wire and polypropylene cover 8.5
₹12,400.00INR / Each
Bel-Art Poxygrid Benchtop Biohazard Bag Stand WW-06496-60 Mfr # H13193-1000
COMPARE
11 Epoxy-coated steel wire 8.5
₹7,500.00INR / Each