Bürkle Industrial Vacuum Pumps for Sealed Liquid Sampling
Durable vacuum-operated liquid sample collection
  • Reach narrow, less-accessible areas with the small-diameter hose Avoid contamination with the easily replaceable suction hose and sample bottle No-hassle maintenance-free sampler
These rugged vacuum samplers are ideal for sampling liquids and filling liquids from a maximum suction depth of 5 m. While sampling, the liquid only comes in contact with suction hose and sample bottle. Avoiding cross-contamination and ensure the authenticity of the sample. The suction hose can quickly and easily be changed as necessary. Built-in venting valve which features fill level control, automatic return flow, and overfill protection.
The aluminum sampler (model 06040-99) is ideal for sampling solvents and flammable liquids in explosion categories IIA, IIB, and IIC. This model comes with an earthing cable and electrically conductive tubing for sampling of these liquids. This sampler can be used with sample bottles 06041-29 or 06041-30 and suction tube 06040-98.
The polypropylene sampler (model 06040-97) is ideal for general liquid sampling of weak acids, alkalis, detergents, and chemical baths. This sampler can be used with the 06040-16 sampling bottle and 06040-98 suction hose.
254,000.00 - ₹329,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Burkle 5305-1000 Compact Industrial Liquid Vacuum Sampler, Polypropylene, Complete Kit; 250 mL 0604097 Mfr # 5305-1000
COMPARE
Small industrial liquid vaccum sampler, PP, complete kit; 250 mL
₹254,000.00INR / Each
Burkle 5305-1100 Small iCompact Industrial Liquid Vacuum Sampler, Stainless Steel, Complete Kit; 250 mL 0604099 Mfr # 5305-1100
COMPARE
Small industrial liquid vaccum sampler, SS, complete kit; 250 mL
₹329,000.00INR / Each