Bürkle Industrial Vaccum Pumps for Sealed Liquid Sampling Accessories
Accessories for the vacuum-operated liquid sample collection
  • Reach narrow, less-accessible areas with the small-diameter hose
  • Avoid contamination with the easily replaceable suction hose and sample bottle
These accessories are suitable for use with the Bürkle industrial vacuum pumps for sealed liquid sampling.
The aluminum sampler (model 06040-99) is ideal for sampling solvents and flammable liquids in explosion categories IIA, IIB, and IIC. This model comes with an earthing cable and electrically conductive tubing for sampling of these liquids. This sampler can be used with sampling bottles 06041-29 or 06041-30 and suction tube 06040-98.
The polypropylene sampler (model 06040-97) is ideal for general liquid sampling of weak acids, alkalis, detergents, and chemical baths. This sampler can be used sampling bottle 06040-16 and suction hose 06040-98.
7,100.00 - ₹8,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Wetted Materials Description
Availability Pricing
Burkle 5314-0050 Sample Bottle, Glass, GL45 cap, Transparent; 500 mL 0604129 Mfr # 5314-0050
COMPARE
Glass Sample bottle, glass, GL45 cap, transparent; 500 mL
₹7,100.00INR / Each
Burkle 5314-0100 Sample Bottle, Glass, GL45 cap, Transparent; 1000 mL 0604130 Mfr # 5314-0100
COMPARE
Glass Sample bottle, glass, GL45 cap, transparent; 1000 mL
₹8,400.00INR / Each