Bürkle Hand-Operated Submersible Sampling Vessel with Rod
Easy single-handed operation of liquid sampling
  • Quickly sample thin to viscous liquid samples
  • Safely and efficiently fill containers with the discharge funnel
  • Constructed of durable 316L stainless steel
Effortlessly sample a wide range of liquids with these single-hand operated sampling vessels. There are two models to choose from. Items 06041-67 and -68 open with a thumb operated draw and release mechanism, emptying the liquid from the discharge funnel. The valve within the funnel is opened with the thumb operated handle. The second model, available as part numbers 06041-69 and -70, features a gravity compression release. The valve is opened by attaching it to a sample container, and gravity forces the liquid out of the discharge vessel.
84,900.00 - ₹93,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description Length (in) Sample Volume
Availability Pricing
Burkle 5326-0001 Fluid sampler, thumb operated draw/release, SS, 46 cm x 32 mm; 50 mL 0604167 Mfr # 5326-0001
COMPARE
Hand-Operated Liquid Sampler, Thumb Operated Draw and Release, Stainless Steel; 50 mL 18.1 50 mL
₹84,900.00INR / Each
Burkle 5326-0002 Fluid sampler, thumb operated draw/release, SS, 54 cm x 32 mm; 100 mL 0604168 Mfr # 5326-0002
COMPARE
Hand-Operated Liquid Sampler, Thumb Operated Draw and Release, Stainless Steel; 100 mL 21.3 100 mL
₹93,700.00INR / Each