B&K Precision Single Output DC Power Supplies
Overload protection against short circuit, reverse polarity and over voltage
These durable power supplies continuously monitor voltage and current output on two analog or digital displays. Operate constant voltage or current output to suit your application. Power supplies operate in parallel and series, feature coarse and fine voltage controls, and full overload protection.
107,000.00 - ₹183,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
B&K Precision 1621A Single Output Power Supply, Digital Display, 0 to 18 VDC, 0 to 5A 2686802 Mfr # 1621A
COMPARE
0 to 5 A 0 to 18 VDC < 0.5 mVrms; < 3 mArms
₹107,000.00INR / Each
B&K Precision 1627A Single Output Power Supply, Digital Display, 0 to 30 VDC, 0 to 3A; Horizontal 2686810 Mfr # 1627A
COMPARE
0 to 3 A 0 to 30 VDC < 0.5 mVrms; < 3 mArms
₹111,000.00INR / Each
B&K Precision 1715A Single Output Power Supply, Digital Display, 0 to 60 VDC, 0 to 2A 2686814 Mfr # 1715A
COMPARE
0 to 2 A 0 to 60 VDC < 1 mVrms; < 3 mArms
₹183,000.00INR / Each
B&K Precision 1735A Single Output Power Supply, Digital Display, 0 to 30 VDC, 0 to 3A; Vertical 2686818 Mfr # 1735A
COMPARE
0 to 3 A 0 to 30 VDC < 1 mVrms; < 3 mArms
₹167,000.00INR / Each