B&K Precision High Power Switching DC Power Supplies
Rotary encoder control allows precise voltage and current setting
  • Automatic constant voltage and constant current crossover operation
  • Save up to 3 user-defined voltage and current presets for quick recall
  • Analog remote control function
  • Remote sensing terminal (model 1900 only)
  • Overvoltage, overtemperature, and overload protection
  • Flexible operating modes: Front panel (normal), preset, or remote control
  • Front panel 5 amp auxiliary output
These switching DC power supplies are designed for a wide range of applications requiring high current including production testing, telecommunications, R&D, service, university labs, and more. Power supplies provide various configurations of high-output voltage or high-output current all in a small, lightweight form factor. The dual-action push-button interface allows you to set both coarse and fine, as well as voltage and current levels, quickly and precisely.
302,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
B&K Precision 1900 DC Switching Power Supply, 960W 2004355 Mfr # 1900B
COMPARE
0 to 60 A 1 to 16 VDC ≤ 5 mVrms, ≤ 50 mVp-p
₹302,000.00INR / Each
B&K Precision 1901 DC Switching Power Supply, 960W 2004356 Mfr # 1901B
COMPARE
0 to 30 A 1 to 32 VDC ≤ 5 mVrms, ≤ 50 mVp-p
₹302,000.00INR / Each
B&K Precision 1902 DC Switching Power Supply, 900W 2004357 Mfr # 1902B
COMPARE
0 to 15 A 1 to 60 VDC ≤ 5 mVrms, ≤ 100 mVp-p
₹302,000.00INR / Each