Aven Tools Carbon Fiber Tweezers
ESD safe – ideal for handling sensitive materials such as electronic components
  • Offers high rigidity, excellent tensile, and flexural strength
  • Chemical resistance to oils, grease, fuels, and non-polar solvents
  • Heat resistance up to 492.8°f (256°C)
These ESD safe tweezers, made of polyetheretherketone (PEEK) reinforced with carbon fiber, offer excellent wear and abrasion resistance. The conductive synthetic fiber tips are ideal for handling sensitive materials and devices such as electronic components, micro-mechanical parts, glass, and ceramic substrates. Each tweezer is manually finished and tested for maximum precision.
11,200.00INR / Pkg of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
ESD Safe Overall Length (in) Style
Availability Pricing
Aven Tools 5-CF Carbon Fiber Tweezer, Straight, Ultra-Fine Tips, 0.5 mm (0.02") Wide WW-07289-00 Mfr # 18762
COMPARE
Yes 5 Straight, Ultra-Fine Tips,
₹11,200.00INR / Pkg of 1
Aven Tools 302-CF Carbon Fiber Tweezer, Straight, Broad Tips, 2.2 mm (0.09") Wide WW-07289-02 Mfr # 18.702
COMPARE
Yes 5 Straight, Broad Tips
₹11,200.00INR / Pkg of 1
Aven Tools 304-CF Carbon Fiber Tweezer, Straight, Ultra-Fine Tips, 0.6 mm (0.024") Wide WW-07289-04 Mfr # 18.704
COMPARE
Yes 5 Straight, Ultra-Fine Tips
₹11,200.00INR / Pkg of 1
Aven Tools 7-CGF Carbon Fiber Tweezers, Curved, Fine Tips, 0.6 mm (0.024") Wide WW-07289-09 Mfr # 18769
COMPARE
Yes 5 Curved, Fine Tips
₹11,200.00INR / Each