Scienceware Autoclavable Biohazard Disposable Bags, HDPE
Indicator patch darkens when sterile
  • Precautionary procedures printed in four languages
  • Meet/exceed ASTM D 1709-85
  • Autoclavable at 250°F (121°C)
These high-molecular weight bags feature an indicator patch that darkens when sterile. Precautionary procedures are printed in English, French, German, and Spanish. Bags are 1.25-mil thick and meet/exceed ASTM D 1709-85.
Optional bag stands are made of aluminum tubing with baked on epoxy coating. The stands fold flat for convenient storage and include a polystyrene safety drip tray that securely supports the bag in the stand. Order stands and plastic ties separately.
27,200.00 - ₹73,500.00INR / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Material Thickness (mil)
Availability Pricing
Scienceware HDPE Biohazard Bags w/ Sterilization Patch, 1 Gallon, 12 x 24"; 100/CS 0649730 Mfr # 13160-0009
COMPARE
1 HDPE 1.25
₹27,200.00INR / Pkg of 100
Scienceware HDPE Biohazard Bags w/ Sterilization Patch, 12 Gallons, 24 x 30"; 100/CS 0649740 Mfr # 13161-0009
COMPARE
12 HDPE 1.25
₹68,300.00INR / Pkg of 100
Scienceware HDPE Biohazard Bags w/ Sterilization Patch, 20 Gallons, 24 x 36"; 100/CS 0649750 Mfr # F 13162-0009
COMPARE
20 HDPE 1.25
₹73,500.00INR / Pkg of 100